Anti-Legionella pneumophila Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIL1663-08EA 414 GBP
USBIL1663-08
Anti-Legionella pneumophila Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 1: 800

Type: Primary
Antigen: Legionella pneumophila
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR