Anti-IRF8 Rabbit Monoclonal Antibody [clone: D20D8]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBII7662-93GEA 520 GBP
USBII7662-93G
Anti-IRF8 Rabbit Monoclonal Antibody [clone: D20D8]
Antibodies
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50ChImmunoprecipitation: 1:25

Type: Primary
Antigen: IRF8 (interferon regulatory factor 8)
Clonality: monoclonal
Clone: D20D8
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Monkey, Mouse, Rat, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR