Anti-IgG1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 8L354]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBII1904-75W1EA 595 GBP
USBII1904-75W1
Anti-IgG1 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 8L354]
Antibodies
Do not freeze!

Type: Primary
Antigen: IgG1
Clonality: monoclonal
Clone: 8L354
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR