Anti-APP Rabbit polyclonal antibody unconjugated

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIA2275-87JEA 652 GBP
USBIA2275-87J
Anti-APP Rabbit polyclonal antibody unconjugated
Antibodies
  • Recommended dilution: ELISA: 0.1-1 μg/ml; Western Blot: 0.2-2μg/ml; Immunofluorescence: 1-5 μg/ml; Immunochemistry: 0.1-1 μg/ml

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR