Anti-COL4 Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI137869EA 539 GBP
USBI137869
Anti-COL4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:5000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR