Anti-ATP1B1 Goat Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI135913EA 594 GBP
USBI135913
Anti-ATP1B1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:28000; Western Blot: 0.1-0. 3?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ATP1B1 (ATPase, Na+/K+ transporting, beta 1 polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR