Anti-PI4KB Mouse Monoclonal Antibody [clone: 3B1]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI131304EA 490 GBP
USBI131304
Anti-PI4KB Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
Type: Primary
Antigen: PI4KB (phosphatidylinositol 4-kinase, catalytic, beta)
Clonality: monoclonal
Clone: 3B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR