Anti-LSS Mouse Monoclonal Antibody [clone: 2E4]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI129243EA 490 GBP
USBI129243
Anti-LSS Mouse Monoclonal Antibody [clone: 2E4]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: LSS (lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase))
Clonality: monoclonal
Clone: 20000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR