Anti-Neurofilament H Mouse Monoclonal Antibody [clone: 13B838]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI031395EA 444 GBP
USBI031395
Anti-Neurofilament H Mouse Monoclonal Antibody [clone: 13B838]
Antibodies
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: Neurofilament H
Clonality: Monoclonal
Clone: 13B838
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Chicken, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR