Anti-Lamin A, C Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI031382EA 412 GBP
USBI031382
Anti-Lamin A, C Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 1:5000; Western Blot: 1:5000

Type: Primary
Antigen: Lamin A, C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mammalian, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR