Anti-RPS6KB2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 164CT21.2.2]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI031228-APCEA 799 GBP
USBI031228-APC
Anti-RPS6KB2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 164CT21.2.2]
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Monoclonal
Clone: 164CT21.2.2
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR