Anti-EPO Mouse Monoclonal Antibody [clone: 13B262]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI030184EA 494 GBP
USBI030184
Anti-EPO Mouse Monoclonal Antibody [clone: 13B262]
Antibodies
Type: Primary
Antigen: EPO (erythropoietin)
Clonality: Monoclonal
Clone: 13B262
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR