Anti-INSIG1 Rabbit Polyclonal Antibody

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI029830EA 466 GBP
USBI029830
Anti-INSIG1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: INSIG1 (insulin induced gene 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR