Anti-Biotin Goat Polyclonal Antibody (Cy5.5®)

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK700-113-098EA 900 GBP
ROCK700-113-098
Anti-Biotin Goat Polyclonal Antibody (Cy5.5®)
Antibodies
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:100-1:1000; Flow Cytometry: 1:200-1:400; Flow Cytometry: 1:200-1:400
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR