Anti-GFP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9F9.F9]

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK600-305-215EA 1580 GBP
ROCK600-305-215
Anti-GFP Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9F9.F9]
Antibodies
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant Proteins. This antibody can be used to detect GFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: AP (Alkaline Phosphatase)
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR