Anti-GST Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK600-106-200EA 1070 GBP
ROCK600-106-200
Anti-GST Goat Polyclonal Antibody (Biotin)
Antibodies
Primary Goat Anti-GLUTATHIONE-S-TRANSFERASE (Schistosoma japonicum) IgG Fraction Reacts with Pan

Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR