Anti-FITC Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK600-106-096EA 987 GBP
ROCK600-106-096
Anti-FITC Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Fluorescein Antibody Biotin Conjugated is suitable for immunoblotting, ELISA, in situ hybridization, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR