Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Supplier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK200-4298EA 5220 GBP
ROCK200-4298
Anti-Biotin Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antibodies
Type: Primary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR