Anti-SV40 T Antigen (Ab-1) (PAb419) Mouse Monoclonal Antibody [clone: PAb419]

Supplier: Oncogene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

DP01-100EA 439 GBP
DP01-100
Anti-SV40 T Antigen (Ab-1) (PAb419) Mouse Monoclonal Antibody [clone: PAb419]
Antibodies
Type: Primary
Antigen: SV40 T Antigen (Ab-1) (PAb419)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR