Anti-Clathrin Heavy Chain (X22) Mouse Monoclonal Antibody

Supplier: Oncogene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CP45-100EA 449 GBP
CP45-100
Anti-Clathrin Heavy Chain (X22) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Clathrin Heavy Chain (X22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR