Anti-CAT Rabbit Polyclonal Antibody

Supplier: Oncogene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

219010-1EA 174 GBP
219010-1
Anti-CAT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CAT (catalase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR