Anti-APOB Goat Polyclonal Antibody

Supplier: Oncogene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

178467-1EA 141 GBP
178467-1
Anti-APOB Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR