Anti-GFP Rabbit polyclonal antibody unconjugated

Supplier: GenScript Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

GENSA01388-40EA 159 GBP
GENSA01388-40
Anti-GFP Rabbit polyclonal antibody unconjugated
Antibodies
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR