Anti-NWSHPQFEK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GenScript Biotech
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

GENSA00875-40EA 159 GBP
GENSA00875-40
Anti-NWSHPQFEK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Tag
Antigen: NWSHPQFEK
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR