Anti-TLR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G6]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNBP2-27251EA 687 GBP
NOVUNBP2-27251
Anti-TLR7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4G6]
Antibodies
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: monoclonal
Clone: 4G6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR