Anti-TP73 Mouse Monoclonal Antibody [clone: 38C674.2]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNBP2-24873EA 580 GBP
NOVUNBP2-24873
Anti-TP73 Mouse Monoclonal Antibody [clone: 38C674.2]
Antibodies
Type: Primary
Antigen: TP73 (tumor protein p73)
Clonality: monoclonal
Clone: 38C674.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR