Anti-MUL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNBP2-24541EA 532 GBP
NOVUNBP2-24541
Anti-MUL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MUL1 (mitochondrial E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR