Anti-TNC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4C8MS]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB110-68136PEEA 723 GBP
NOVUNB110-68136PE
Anti-TNC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4C8MS]
Antibodies
Type: Primary
Antigen: TNC (tenascin C)
Clonality: monoclonal
Clone: 4C8MS
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR