Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 755) [clone: VG76e]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB100-648IREA 769 GBP
NOVUNB100-648IR
Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 755) [clone: VG76e]
Antibodies
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG76e
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR