Anti-Livin Mouse Monoclonal Antibody [clone: 88C570]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB100-56548EA 586 GBP
NOVUNB100-56548
Anti-Livin Mouse Monoclonal Antibody [clone: 88C570]
Antibodies
Type: Primary
Antigen: Livin
Clonality: monoclonal
Clone: 88C570
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR