Anti-CYC Mouse Monoclonal Antibody [clone: 7H8.2C12]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB100-56503EA 620 GBP
NOVUNB100-56503
Anti-CYC Mouse Monoclonal Antibody [clone: 7H8.2C12]
Antibodies
Type: Primary
Antigen: CYC (cytochrome C)
Clonality: monoclonal
Clone: 7H8.2C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR