Anti-Cre Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NOVUNB100-56134-50EA 565 GBP
NOVUNB100-56134-50
Anti-Cre Rabbit Polyclonal Antibody
Antibodies
The Cre Antibody from Novus Biologicals is a rabbit polyclonal antibody to Cre. This antibody reacts with cre/lox system. The Cre Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page, Immunohistochemistry Free-Floating.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:1000-1:5000

Type: Primary
Antigen: Cre
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cre/lox System
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR