Anti-IgG Chicken Polyclonal Antibody (Biotin)

Supplier: MARKET SOURCE PART PROCESS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

MSPPAHIGGFAB-BEA 0 GBP
MSPPAHIGGFAB-B
Anti-IgG Chicken Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR