Anti-TRAF2 Rabbit Monoclonal Antibody [clone: EPR6048]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB126758-100EA 380 GBP
ABCAAB126758-100
Anti-TRAF2 Rabbit Monoclonal Antibody [clone: EPR6048]
Antibodies
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: monoclonal
Clone: EPR6048
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR