Anti-IgG2a Goat Polyclonal Antibody

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB98694-500EA 170 GBP
ABCAAB98694-500
Anti-IgG2a Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR