Anti-GFAP Rabbit monoclonal antibody unconjugated [clone: EPR1034Y]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB68428-100EA 485 GBP
ABCAAB68428-100
Anti-GFAP Rabbit monoclonal antibody unconjugated [clone: EPR1034Y]
Antibodies
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone: EPR1034Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR