Anti-SYP Rabbit Monoclonal Antibody [clone: EP1098Y]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB52636-100EA 350 GBP
ABCAAB52636-100
Anti-SYP Rabbit Monoclonal Antibody [clone: EP1098Y]
Antibodies
Type: Primary
Antigen: SYP (synaptophysin)
Clonality: monoclonal
Clone: EP1098Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR