Anti-S100A12 Goat Polyclonal Antibody

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB36001-100EA 405 GBP
ABCAAB36001-100
Anti-S100A12 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: S100A12 (S100 calcium binding protein A12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR