Anti-MYC Rabbit Monoclonal Antibody [clone: Y69]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB32072-100EA 420 GBP
ABCAAB32072-100
Anti-MYC Rabbit Monoclonal Antibody [clone: Y69]
Antibodies
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone: Y69
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR