Anti-CARD8 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABCAAB24186-100EA 525 GBP
ABCAAB24186-100
Anti-CARD8 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CARD8 (caspase recruitment domain family, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR