Anti-POLR2A Rabbit Polyclonal Antibody

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB127542-100 ORB127542-200
BIRBORB127542-100EA 0 GBP
BIRBORB127542-100 BIRBORB127542-200
Anti-POLR2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POLR2A (polymerase (RNA) II (DNA directed) polypeptide A, 220kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR