Anti-MYC Mouse Monoclonal Antibody [clone: OTI1A6]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA500002EA 583 GBP
ORIGTA500002
Anti-MYC Mouse Monoclonal Antibody [clone: OTI1A6]
Antibodies
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR