Anti-LGR5 Mouse Antibody

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA400001EA 589 GBP
ORIGTA400001
Anti-LGR5 Mouse Antibody
Antibodies
Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR