Anti-GPX2 Goat Polyclonal Antibody

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA303039EA 513 GBP
ORIGTA303039
Anti-GPX2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: GPX2 (glutathione peroxidase 2 (gastrointestinal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR