Anti-Myc Tag None Monoclonal Antibody [clone: 9E10]

Supplier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORIGTA150121EA 466 GBP
ORIGTA150121
Anti-Myc Tag None Monoclonal Antibody [clone: 9E10]
Antibodies
Type: Tag
Antigen: Myc Tag (Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR