Anti-PLXND1 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP3584AEA 339 GBP
ABGEAP3584A
Anti-PLXND1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLXND1 (plexin D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR