Anti-ARFGAP1 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP2303BEA 339 GBP
ABGEAP2303B
Anti-ARFGAP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARFGAP1 (ADP-ribosylation factor GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR