Anti-ALDH1L2 Rabbit Polyclonal Antibody [clone: 164CT21.2.2]

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABGEAP11288BEA 339 GBP
ABGEAP11288B
Anti-ALDH1L2 Rabbit Polyclonal Antibody [clone: 164CT21.2.2]
Antibodies
Type: Primary
Antigen: ALDH1L2 (aldehyde dehydrogenase 1 family, member L2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR