Anti-BMI1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AVIVARP34213_P050EA 339 GBP
AVIVARP34213_P050
Anti-BMI1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR