Anti-SOX18 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AVIVARP33056_P050EA 339 GBP
AVIVARP33056_P050
Anti-SOX18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOX18 (SRY (sex determining region Y)-box 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR